Gene Transcripts

Gene Symbol
Jakmip2
Entrez Gene ID
76217
Gene Name
janus kinase and microtubule interacting protein 2
Gene Aliases
6430702L21Rik, AI850334, D930046L20Rik
Chromosome Location
Chr. 18: 43527096 - 43687773 on Build GRCm38
UniGene ID
Mm.134602
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163637.1 NP_001157109.1 7 1584
GenBank mRNA AK133750.1 - 7 1549
AK139194.1 - 2 2427
BC094439.1 - 6 1146

View Full Product Details