Gene Transcripts
- Gene Symbol
- Maf
- Entrez Gene ID
- 54267
- Gene Name
- v-maf avian musculoaponeurotic fibrosarcoma oncogene homolog
- Gene Aliases
- Maf2, cMaf
- Chromosome Location
- Chr. 19: 48179826 - 48196754 on Build Rnor_6.0
- UniGene ID
- Rn.10726
- Species
- Rattus norvegicus
- Gene Symbol
- Mafa
- Entrez Gene ID
- 366949
- Gene Name
- v-maf avian musculoaponeurotic fibrosarcoma oncogene A
- Gene Aliases
- RGD1562627
- Chromosome Location
- Chr. 7: 116779368 - 116781815 on Build Rnor_6.0
- Species
- Rattus norvegicus
View Full Product Details