Gene Transcripts

Gene Symbol
Plvap
Entrez Gene ID
56765
Gene Name
plasmalemma vesicle associated protein
Gene Aliases
Pv1
Chromosome Location
Chr. 16: 19906354 - 19918668 on Build Rnor_6.0
UniGene ID
Rn.53915
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020086.1 NP_064471.1 1, 2 390
GenBank mRNA AF154831.1 - 1, 2 390
BC070900.1 - 1, 2 390 7100403

View Full Product Details