Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199864.1 | NP_001186793.1 | 3 | 312 | |
GenBank mRNA | BC011637.1 | - | 3 | 248 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199839.1 | NP_001186768.1 | 3 | 267 | |
NM_004050.4 | NP_004041.1 | 3 | 312 | ||
GenBank mRNA | AK289519.1 | - | 3 | 267 | |
BC021198.2 | - | 2 | 249 | 3944307 | |
BC104789.1 | - | 3 | 248 | 8143792 | |
BC113522.1 | - | 3 | 248 | 8322574 | |
BP216712.1 | - | 3 | 312 | ||
BT019549.1 | - | 1 | 83 | ||
D87461.1 | - | 3 | 259 | ||
DA196433.1 | - | 3 | 271 | ||
DA288957.1 | - | 3 | 267 | ||
DA451936.1 | - | 3 | 224 | ||
U59747.1 | - | 1 | 83 |