Gene Transcripts

Gene Symbol
Ccdc80
Entrez Gene ID
64387
Gene Name
coiled-coil domain containing 80
Gene Aliases
CL2, Dro1, Ssg1
Chromosome Location
Chr. 11: 60645660 - 60679249 on Build Rnor_6.0
UniGene ID
Rn.2193
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022543.2 NP_071988.2 2 1887
GenBank mRNA AF223677.1 - 2 1994
AY548105.1 - 2 1887
BC070926.1 - 1 1225

View Full Product Details