Gene Transcripts

Gene Symbol
Six3
Entrez Gene ID
78974
Gene Name
SIX homeobox 3
Gene Aliases
Chromosome Location
Chr. 6: 8886730 - 8891094 on Build Rnor_6.0
UniGene ID
Rn.123641
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023990.1 NP_076480.1 1 758
GenBank mRNA AB030833.1 - 1 758

View Full Product Details