Gene Transcripts

Gene Symbol
Alas1
Entrez Gene ID
65155
Gene Name
5'-aminolevulinate synthase 1
Gene Aliases
ALAS
Chromosome Location
Chr. 8: 114927704 - 114941038 on Build Rnor_6.0
UniGene ID
Rn.97126
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024484.2 NP_077810.2 6 939
GenBank mRNA BC061793.1 - 6 939
J03190.1 - 5 863
J04044.1 - 5 870

View Full Product Details