Gene Transcripts

Gene Symbol
Fcna
Entrez Gene ID
83517
Gene Name
ficolin A
Gene Aliases
Fcn1
Chromosome Location
Chr. 3: 2850006 - 2853230 on Build Rnor_6.0
UniGene ID
Rn.20051
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031348.2 NP_112638.2 10 1079
XM_008761614.1 XP_008759836.1 10 1112
GenBank mRNA AB026057.2 - 10 990
BC088128.1 - 10 1079 7376326

View Full Product Details