Gene Transcripts

Gene Symbol
Cpz
Entrez Gene ID
83575
Gene Name
carboxypeptidase Z
Gene Aliases
Chromosome Location
Chr. 14: 80402954 - 80426203 on Build Rnor_6.0
UniGene ID
Rn.11056
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031766.1 NP_113954.1 4 740
XM_006251151.2 XP_006251213.1 3 735
GenBank mRNA AF017637.1 - 4 740
AF017638.1 - 4 740

View Full Product Details