Gene Transcripts

Gene Symbol
Pafah1b1
Entrez Gene ID
83572
Gene Name
platelet-activating factor acetylhydrolase 1b, regulatory subunit 1
Gene Aliases
LIS1
Chromosome Location
Chr. 10: 61457216 - 61498601 on Build Rnor_6.0
UniGene ID
Rn.5827
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031763.3 NP_113951.1 1397
GenBank mRNA AF016049.1 - 1212
BC072510.1 - 1397 7104733
FQ224997.1 - 1122

View Full Product Details