Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033236.1 | NP_150239.1 | 7 | 569 | |
GenBank mRNA | BC061542.1 | - | 7 | 611 | |
D50694.1 | - | 7 | 569 | ||
FQ209873.1 | - | 4 | 372 | ||
FQ212821.1 | - | 7 | 599 | ||
FQ213265.1 | - | 4 | 372 | ||
FQ215767.1 | - | 4 | 372 | ||
FQ225383.1 | - | 4 | 372 | ||
FQ230973.1 | - | 7 | 628 | ||
FQ234824.1 | - | 4 | 372 |