Gene Transcripts

Gene Symbol
Kap
Entrez Gene ID
24937
Gene Name
kidney androgen regulated protein
Gene Aliases
Chromosome Location
Chr. 4: 167549922 - 167552973 on Build Rnor_6.0
UniGene ID
Rn.11143
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052802.1 NP_434689.1 2 336
XM_006237483.2 XP_006237545.1 2 264
GenBank mRNA U25808.1 - 2 336

View Full Product Details