Gene Transcripts

Gene Symbol
Slc7a8
Entrez Gene ID
84551
Gene Name
solute carrier family 7 (amino acid transporter light chain, L system), member 8
Gene Aliases
Lat2, Lat4
Chromosome Location
Chr. 15: 33369245 - 33428942 on Build Rnor_6.0
UniGene ID
Rn.82734
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053442.1 NP_445894.1 2 924
GenBank mRNA AB024400.1 - 2 924

View Full Product Details