Gene Transcripts

Gene Symbol
Cox4i2
Entrez Gene ID
84683
Gene Name
cytochrome c oxidase subunit IV isoform 2 (lung)
Gene Aliases
Cox4b, CoxIV-2
Chromosome Location
Chr. 3: 148234546 - 148245424 on Build Rnor_6.0
UniGene ID
Rn.7214
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053472.1 NP_445924.1 5 426
XM_006235280.2 XP_006235342.1 5 781
GenBank mRNA AF255347.1 - 5 426

View Full Product Details