Gene Transcripts

Gene Symbol
Ftcd
Entrez Gene ID
89833
Gene Name
formimidoyltransferase cyclodeaminase
Gene Aliases
Chromosome Location
Chr. 20: 12806957 - 12820466 on Build Rnor_6.0
UniGene ID
Rn.20140
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053567.1 NP_446019.1 3, 4 380
GenBank mRNA AF079233.3 - 3, 4 380
BC091134.1 - 3, 4 392 7369085

View Full Product Details