Gene Transcripts

Gene Symbol
Tm4sf4
Entrez Gene ID
116467
Gene Name
transmembrane 4 L six family member 4
Gene Aliases
Lrtm4
Chromosome Location
Chr. 2: 147532032 - 147547254 on Build Rnor_6.0
UniGene ID
Rn.13425
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053785.1 NP_446237.1 1 439
GenBank mRNA AF205717.1 - 1 439
BC091131.1 - 1 254 7324973

View Full Product Details