Gene Transcripts

Gene Symbol
Csf2rb
Entrez Gene ID
171081
Gene Name
colony stimulating factor 2 receptor, beta, low-affinity (granulocyte-macrophage)
Gene Aliases
Csf2rb1
Chromosome Location
Chr. 7: 119554383 - 119568248 on Build Rnor_6.0
UniGene ID
Rn.42930
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133555.1 NP_598239.1 1, 2 251
GenBank mRNA AJ000555.1 - 1, 2 251
S79263.1 - 1, 2 251

View Full Product Details