Gene Transcripts

Gene Symbol
Eif2b3
Entrez Gene ID
171145
Gene Name
eukaryotic translation initiation factor 2B, subunit 3
Gene Aliases
Chromosome Location
Chr. 5: 135882955 - 135949276 on Build Rnor_6.0
UniGene ID
Rn.10577
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133609.2 NP_598293.2 3 299
XM_006238630.2 XP_006238692.1 3 270
GenBank mRNA BC072507.1 - 3 299
U38253.1 - 3 279

View Full Product Details