Gene Transcripts

Gene Symbol
Ppp1r14d
Entrez Gene ID
259226
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 14D
Gene Aliases
Gbpi
Chromosome Location
Chr. 3: 111022920 - 111037166 on Build Rnor_6.0
UniGene ID
Rn.162687
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172011.1 NP_742008.1 1 142
GenBank mRNA AY122322.1 - 1 142

View Full Product Details