Gene Transcripts
- Gene Symbol
- Fxyd1
- Entrez Gene ID
- 58971
- Gene Name
- FXYD domain-containing ion transport regulator 1
- Gene Aliases
- Plm
- Chromosome Location
- Chr. 1: 89484197 - 89488278 on Build Rnor_6.0
- UniGene ID
- Rn.3828
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031648.2 |
NP_113836.2
|
3, 4 |
173 |
|
XM_006228838.2 |
XP_006228900.1
|
4, 5 |
393 |
|
XM_006228839.2 |
XP_006228901.1
|
3, 4 |
168 |
|
XM_008759184.1 |
XP_008757406.1
|
3, 4 |
182 |
|
GenBank mRNA |
CB712570.1 |
- |
3, 4 |
173 |
|
FM109156.1 |
- |
3, 4 |
127 |
|
FQ211349.1 |
- |
3, 4 |
184 |
|
FQ224131.1 |
- |
3, 4 |
136 |
|
FQ224338.1 |
- |
3, 4 |
219 |
|
U72246.1 |
- |
2, 3 |
87 |
|
View Full Product Details