Gene Transcripts

Gene Symbol
Fgf3
Entrez Gene ID
170633
Gene Name
fibroblast growth factor 3
Gene Aliases
Int2
Chromosome Location
Chr. 1: 218003018 - 218006942 on Build Rnor_6.0
UniGene ID
Rn.81236
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130817.1 NP_570830.1 1 149
GenBank mRNA AB079262.1 - 1 149

View Full Product Details