Gene Transcripts

Gene Symbol
Bmp7
Entrez Gene ID
12162
Gene Name
bone morphogenetic protein 7
Gene Aliases
OP1
Chromosome Location
Chr. 2: 172868012 - 172940321 on Build GRCm38
UniGene ID
Mm.392150
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007557.3 NP_031583.2 2 781
GenBank mRNA AK020411.1 - 2 553
BC010771.1 - 2 753 4218495
X56906.1 - 2 632

View Full Product Details