Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_008161.3 | NP_032187.2 | 2 | 273 | |
GenBank mRNA | AK002219.1 | - | 2 | 202 | |
AK002262.1 | - | 2 | 202 | ||
AK004942.1 | - | 2 | 202 | ||
AK146760.1 | - | 2 | 202 | ||
BC003339.1 | - | 2 | 185 | 3257333 | |
BC037027.1 | - | 2 | 220 | 5370640 | |
BC037134.1 | - | 2 | 199 | ||
BC049235.1 | - | 2 | 189 | 4221025 | |
BC061950.1 | - | 2 | 177 | 6528998 | |
BY100863.1 | - | 2 | 275 | ||
BY339672.1 | - | 2 | 275 | ||
CB601893.1 | - | 2 | 205 | ||
U13705.1 | - | 2 | 177 |