Gene Transcripts

Gene Symbol
Ifngr2
Entrez Gene ID
15980
Gene Name
interferon gamma receptor 2
Gene Aliases
Ifgr2, Ifgt
Chromosome Location
Chr. 16: 91547094 - 91564007 on Build GRCm38
UniGene ID
Mm.249364
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008338.3 NP_032364.1 2 184
GenBank mRNA AK086929.1 - 2 198
AK137165.1 - 2 207
AK143214.1 - 2 199
AK146995.1 - 202
AK165193.1 - 2 200
AK169907.1 - 2 204
BC055745.1 - 2 184 6403454
S69336.1 - 2 208

View Full Product Details