Gene Transcripts

Gene Symbol
Marcks
Entrez Gene ID
17118
Gene Name
myristoylated alanine rich protein kinase C substrate
Gene Aliases
Macs, PKCSL
Chromosome Location
Chr. 10: 37133243 - 37138926 on Build GRCm38
UniGene ID
Mm.30059
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008538.2 NP_032564.1 2 1393
GenBank mRNA BC046601.1 - 2 1354
M60474.1 - 2 1382
X59374.1 - 2 1284

View Full Product Details