Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021959.2 | NP_068778.1 | 3 | 1198 | |
XM_006715174.3 | XP_006715237.1 | 4 | 1155 | ||
GenBank mRNA | AF070599.1 | - | 2 | 1257 | |
BC003144.1 | - | 4 | 1193 | 2823116 | |
X81003.1 | - | 3 | 1141 |