Gene Transcripts
- Gene Symbol
- Mitf
- Entrez Gene ID
- 17342
- Gene Name
- microphthalmia-associated transcription factor
- Gene Aliases
- BCC2, Bhlhe32, Gsfbcc2, Vitiligo, Wh, bw, mi, vit
- Chromosome Location
- Chr. 6: 97807003 - 98021360 on Build GRCm38
- UniGene ID
- Mm.333284
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001113198.1 |
NP_001106669.1
|
3, 4 |
716 |
|
NM_001178049.1 |
NP_001171520.1
|
3, 4 |
603 |
|
NM_008601.3 |
NP_032627.1
|
2, 3 |
407 |
|
GenBank mRNA |
AB221657.1 |
- |
2, 3 |
269 |
|
AK052895.1 |
- |
3, 4 |
575 |
|
AK154189.1 |
- |
3, 4 |
717 |
|
AK155350.1 |
- |
3, 4 |
717 |
|
AK200280.1 |
- |
1, 2 |
73 |
|
BC108976.2 |
- |
2, 3 |
371 |
40047440 |
BC108977.1 |
- |
2, 3 |
371 |
40047441 |
Z23066.1 |
- |
2, 3 |
398 |
|
View Full Product Details