Gene Transcripts
- Gene Symbol
- Ntsr2
- Entrez Gene ID
- 18217
- Gene Name
- neurotensin receptor 2
- Gene Aliases
- NT2R, NTRL
- Chromosome Location
- Chr. 12: 16653452 - 16660236 on Build GRCm38
- UniGene ID
- Mm.281715
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB041827.1 |
- |
4 |
1427 |
|
U51908.1 |
- |
4 |
1450 |
|
View Full Product Details