Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009077.2 | NP_033103.2 | 3 | 223 | |
GenBank mRNA | AK009044.1 | - | 3 | 194 | |
AK010510.1 | - | 3 | 191 | ||
AK012580.1 | - | 3 | 191 | ||
AK145649.1 | - | 3 | 224 | ||
AK151667.1 | - | 3 | 193 | ||
AK153172.1 | - | 3 | 191 | ||
AK166803.1 | - | 3 | 193 | ||
AK168288.1 | - | 3 | 191 | ||
AK169659.1 | - | 3 | 191 | ||
BC072558.1 | - | 2532 | |||
BC081468.1 | - | 3 | 181 | 6823249 | |
BC082290.1 | - | 3 | 177 | 6485569 | |
BC091732.1 | - | 2 | 169 | 2655484 | |
DQ403025.1 | - | 2 | 88 | ||
L04128.1 | - | 2 | 183 |