Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009056.2 | NP_033082.1 | 11 | 1333 | |
NM_027787.1 | NP_082063.1 | 12 | 1408 | ||
GenBank mRNA | AK044439.1 | - | 12 | 1408 | |
BC004654.1 | - | 11 | 1350 | 3497910 | |
X76089.1 | - | 11 | 1288 |