Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009274.2 | NP_033300.2 | 2, 3 | 405 | |
GenBank mRNA | AB006036.1 | - | 2, 3 | 198 | |
AK014004.1 | - | 3, 4 | 324 | ||
AK136861.1 | - | 2, 3 | 370 | ||
AK161384.1 | - | 3, 4 | 464 | ||
AK169580.1 | - | 2, 3 | 378 | ||
BC020178.1 | - | 2, 3 | 302 | 4507346 | |
BC062941.1 | - | 3, 4 | 350 | ||
U92456.1 | - | 3, 4 | 327 |