Gene Transcripts

Gene Symbol
BCL2A1
Entrez Gene ID
597
Gene Name
BCL2 related protein A1
Gene Aliases
ACC-1, ACC-2, ACC1, ACC2, BCL2L5, BFL1, GRS, HBPA1
Chromosome Location
Chr. 15: 79960890 - 79971301 on Build GRCh38
UniGene ID
Hs.227817
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001114735.1 NP_001108207.1 1 581
NM_004049.3 NP_004040.1 1 581
GenBank mRNA AL110097.1 - 1 449
AY234180.1 - 1 407
BC016281.1 - 1 459 3920808
BG198875.1 - 1 728
BG216703.1 - 1 582
BT007103.1 - 1 399
CD640106.1 - 1 347
CR541937.1 - 1 399
CR541962.1 - 1 399
U27467.1 - 1 433
U29680.1 - 1 477
Y09397.1 - 1 501

View Full Product Details