Gene Transcripts
- Gene Symbol
- ADGRE5
- Entrez Gene ID
- 976
- Gene Name
- adhesion G protein-coupled receptor E5
- Gene Aliases
- CD97, TM7LN1
- Chromosome Location
- Chr. 19: 14381144 - 14408725 on Build GRCh38
- UniGene ID
- Hs.466039
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001025160.2 |
NP_001020331.1
|
8, 9 |
1126 |
|
NM_001784.4 |
NP_001775.2
|
7, 8 |
994 |
|
NM_078481.3 |
NP_510966.1
|
9, 10 |
1273 |
|
XM_011528451.2 |
XP_011526753.1
|
9, 10 |
1225 |
|
XM_017027547.1 |
XP_016883036.1
|
10, 11 |
1137 |
|
GenBank mRNA |
AK097264.1 |
- |
7, 8 |
589 |
|
AK225655.1 |
- |
8, 9 |
835 |
|
AK225677.1 |
- |
7 |
694 |
|
AK292159.1 |
- |
7, 8 |
981 |
|
AK300343.1 |
- |
8, 9 |
826 |
|
AK304553.1 |
- |
5, 6 |
433 |
|
AK308368.1 |
- |
7, 8 |
800 |
|
AK314697.1 |
- |
9, 10 |
973 |
|
AU117846.1 |
- |
6, 7 |
584 |
|
AX775811.1 |
- |
7, 8 |
702 |
|
AX775813.1 |
- |
9, 10 |
941 |
|
AY545551.1 |
- |
|
882 |
|
BC026690.2 |
- |
7, 8 |
690 |
3847171 |
U76764.1 |
- |
9, 10 |
941 |
|
X84700.1 |
- |
7, 8 |
684 |
|
View Full Product Details