Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010577.3 | NP_034707.3 | 2 | 485 | |
GenBank mRNA | AK144921.1 | - | 2 | 271 | |
AK148138.1 | - | 2 | 325 | ||
AK152594.1 | - | 2 | 283 | ||
BC050943.1 | - | 2 | 501 | 6821577 | |
BC053431.1 | - | 3 | 546 | ||
BC059829.1 | - | 2 | 501 | 6821577 | |
X79003.1 | - | 2 | 442 |