Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011876.3 | NP_036006.1 | 3 | 397 | |
GenBank mRNA | AK002699.1 | - | 2 | 312 | |
AK003997.1 | - | 2 | 292 | ||
AK084026.1 | - | 3 | 397 | ||
AK087422.1 | - | 2 | 312 | ||
AK155204.1 | - | 3 | 380 | ||
AK157886.1 | - | 3 | 342 | ||
AK166900.1 | - | 3 | 365 | ||
AK170441.1 | - | 3 | 345 | ||
AY267189.1 | - | 3 | 245 | ||
BC003338.1 | - | 3 | 344 | 3257230 | |
Y17808.1 | - | 3 | 262 |