Gene Transcripts

Gene Symbol
Mpdu1
Entrez Gene ID
24070
Gene Name
mannose-P-dolichol utilization defect 1
Gene Aliases
LEC35, SL15, Supl15h
Chromosome Location
Chr. 11: 69656697 - 69662649 on Build GRCm38
UniGene ID
Mm.89579
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301710.1 NP_001288639.1 5 879
NM_001301711.1 NP_001288640.1 7 1119
NM_011900.4 NP_036030.2 7 1084
GenBank mRNA AB024713.1 - 7 1012
AK080233.1 - 7 1078
AK089426.1 - 7 1026
AK172020.1 - 7 1045
BC026776.1 - 7 1006 4159355
BC048871.1 - 7 1012 5364408

View Full Product Details