Gene Transcripts
- Gene Symbol
- Dmtn
- Entrez Gene ID
- 13829
- Gene Name
- dematin actin binding protein
- Gene Aliases
- AI325486, Epb4.9, Epb49, dematin
- Chromosome Location
- Chr. 14: 70602184 - 70636048 on Build GRCm38
- UniGene ID
- Mm.210863
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001252662.1 |
NP_001239591.1
|
9, 10 |
1216 |
|
NM_001252663.1 |
NP_001239592.1
|
9, 10 |
1216 |
|
NM_001252664.1 |
NP_001239593.1
|
8, 9 |
1043 |
|
NM_001252665.1 |
NP_001239594.1
|
8, 9 |
942 |
|
NM_001252666.1 |
NP_001239595.1
|
7, 8 |
975 |
|
NM_013514.4 |
NP_038542.1
|
10, 11 |
1157 |
|
GenBank mRNA |
AF079846.3 |
- |
8, 9 |
886 |
|
AF155547.1 |
- |
8, 9 |
886 |
|
AK143840.1 |
- |
9, 10 |
1210 |
|
AK149301.1 |
- |
8, 9 |
853 |
|
AK155927.1 |
- |
9, 10 |
1211 |
|
AK158744.1 |
- |
8, 9 |
974 |
|
AK162866.1 |
- |
8, 9 |
894 |
|
AK165204.1 |
- |
|
1209 |
|
AK165294.1 |
- |
9, 10 |
1208 |
|
BC016897.1 |
- |
10, 11 |
1157 |
4240747 |
BC037021.1 |
- |
7, 8 |
962 |
5366040 |
View Full Product Details