Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013684.3 | NP_038712.3 | 7, 8 | 1126 | |
GenBank mRNA | AK085037.1 | - | 7, 8 | 1127 | |
BC012685.1 | - | 9, 10 | 1362 | 3498542 | |
BC016476.1 | - | 6, 7 | 936 | 3584341 | |
BC050136.1 | - | 7, 8 | 1077 | 30020603 | |
D01034.1 | - | 6, 7 | 943 | ||
DQ906043.1 | - | 124 | |||
U63933.1 | - | 8, 9 | 1132 |