Gene Transcripts

Gene Symbol
Dpf1
Entrez Gene ID
29861
Gene Name
D4, zinc and double PHD fingers family 1
Gene Aliases
Neud4
Chromosome Location
Chr. 7: 29303959 - 29317592 on Build GRCm38
UniGene ID
Mm.101885
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013874.2 NP_038902.1 3 323
GenBank mRNA AK142419.1 - 3 302
BC052348.1 - 3 352 6587002
U48238.1 - 3 322

View Full Product Details