Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016772.1 | NP_058052.1 | 2 | 172 | |
GenBank mRNA | AF030343.1 | - | 2 | 172 | |
AK088495.1 | - | 2 | 160 | ||
BC068112.1 | - | 2 | 172 | 5710971 | |
BC087924.1 | - | 2 | 233 | 6706361 |