Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016891.3 | NP_058587.1 | 4, 5 | 549 | |
GenBank mRNA | AB021743.1 | - | 4, 5 | 497 | |
AK054239.1 | - | 4, 5 | 509 | ||
AK078135.1 | - | 4, 5 | 528 | ||
AK088776.1 | - | 4, 5 | 521 | ||
AK190426.1 | - | 1, 2 | 68 | ||
AK217851.1 | - | 1 | 68 | ||
BC006606.1 | - | 4, 5 | 549 | 3157293 | |
BC052678.1 | - | 4, 5 | 545 | 30062580 |