Gene Transcripts

Gene Symbol
Clptm1
Entrez Gene ID
56457
Gene Name
cleft lip and palate associated transmembrane protein 1
Gene Aliases
HS9, N14
Chromosome Location
Chr. 7: 19631580 - 19665030 on Build GRCm38
UniGene ID
Mm.290960
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019649.2 NP_062623.2 14 2010
GenBank mRNA AK148080.1 - 14 1992
AK165742.1 - 13 2704
AK171901.1 - 13 2655
BC018389.1 - 4 596
BC022172.1 - 14 2001 4193279
D67067.1 - 14 1996

View Full Product Details