Gene Transcripts

Gene Symbol
Cend1
Entrez Gene ID
57754
Gene Name
cell cycle exit and neuronal differentiation 1
Gene Aliases
1500001H12Rik, AI415214, BM88
Chromosome Location
Chr. 7: 141426446 - 141429538 on Build GRCm38
UniGene ID
Mm.87027
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021316.4 NP_067291.1 2 408
GenBank mRNA AF243130.1 - 2 346
AK005092.1 - 2 383
AK079045.1 - 2 409
BC023032.1 - 2 346 5366891

View Full Product Details