Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_021381.3 | NP_067356.2 | 2, 3 | 739 | |
GenBank mRNA | AF236082.1 | - | 1, 2 | 493 | |
AF487278.1 | - | 1, 2 | 493 | ||
AK134279.1 | - | 2, 3 | 753 | ||
AK156734.1 | - | 2, 3 | 913 | ||
BC059003.1 | - | 2, 3 | 739 | 6848525 |