Gene Transcripts

Gene Symbol
Jmy
Entrez Gene ID
57748
Gene Name
junction-mediating and regulatory protein
Gene Aliases
Chromosome Location
Chr. 13: 93430097 - 93499808 on Build GRCm38
UniGene ID
Mm.100273
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021310.3 NP_067285.2 10 3306
GenBank mRNA AB221690.1 - 10 2804
AF201390.1 - 10 2930
BC090835.1 - 10 3187 5705427

View Full Product Details