Gene Transcripts

Gene Symbol
Treh
Entrez Gene ID
58866
Gene Name
trehalase (brush-border membrane glycoprotein)
Gene Aliases
2210412M19Rik
Chromosome Location
Chr. 9: 44673233 - 44686305 on Build GRCm38
UniGene ID
Mm.45380
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277847.1 NP_001264776.1 7 730
NM_001277851.1 NP_001264780.1 7 734
NM_021481.3 NP_067456.1 8 835
GenBank mRNA AF136944.1 - 8 830
AK002566.1 - 7 733
AK008912.1 - 8 820
BC019214.1 - 7 725 5064708

View Full Product Details