Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023799.3 | NP_076288.1 | 1, 2 | 368 | |
GenBank mRNA | AF132214.1 | - | 1, 2 | 213 | |
AK012695.1 | - | 2, 3 | 355 | ||
AK088774.1 | - | 1, 2 | 631 | ||
AK129188.1 | - | 1, 2 | 597 | ||
AK145227.1 | - | 1, 2 | 618 | ||
BC020093.1 | - | 1, 2 | 305 | ||
BC041109.1 | - | 1, 2 | 698 | ||
BC054821.1 | - | 1, 2 | 368 | 6515250 |