Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023873.3 | NP_076362.2 | 3, 4 | 257 | |
GenBank mRNA | AF242319.1 | - | 3, 4 | 293 | |
AK020174.1 | - | 3, 4 | 595 | ||
AK020622.1 | - | 3, 4 | 334 | ||
AK021095.1 | - | 3, 4 | 388 | ||
AK049326.1 | - | 3, 4 | 265 | ||
AK133048.1 | - | 3, 4 | 263 | ||
BC050819.1 | - | 3, 4 | 249 | 6517145 | |
BC071260.1 | - | 3, 4 | 257 | 6336351 |