Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025458.2 | NP_079734.1 | 2 | 317 | |
GenBank mRNA | AK007320.1 | - | 2 | 313 | |
AK007518.1 | - | 2 | 318 | ||
AK007830.1 | - | 2 | 311 | ||
AK007885.1 | - | 2 | 318 | ||
BC086923.1 | - | 2 | 317 | 6760985 |