Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026164.2 | NP_080440.2 | 2 | 578 | |
GenBank mRNA | AB044139.1 | - | 2 | 457 | |
AK004621.1 | - | 2 | 545 | ||
AK029616.1 | - | 2 | 544 | ||
AK145776.1 | - | 2 | 488 | ||
AK163211.1 | - | 2 | 546 | ||
AK168475.1 | - | 2 | 541 | ||
BC127056.1 | - | 1 | 438 | 40109753 |